missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PowerEase™ Touch 120W Power Supply and XCell SureLock™ Mini-Cell Welcome Pack

Catalog No. PSC120S
Change view
Click to view available options
Quantity:
1 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PSC120S 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PSC120S Supplier Invitrogen™ Supplier No. PSC120S
  • ONLINE EXCLUSIVE PRICE
    $2,145.00
    $1,394.25 / Each

    Save $750.75

    35.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The PowerEase Touch 120W Power Supply and XCell SureLock Mini-Cell Welcome Pack contains: PowerEase Touch 120W Power Supply and XCell SureLock Mini-Cell

The PowerEase Touch 120W Power Supply and XCell SureLock Mini-Cell Welcome Pack contains:

  • PowerEase Touch 120W Power Supply
  • XCell SureLock Mini-Cell

About the PowerEase Touch 120W Power Supply

The Invitrogen PowerEase Touch 120W Power Supply is a medium-throughput power supply with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for DNA/RNA electrophoresis, SDS-PAGE, and native PAGE. The PowerEase Touch 120W Power Supply offers four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency.

About the XCell SureLock Mini-Cell

The Invitrogen XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Rather than using clamps, the system incorporates the use of a gel tension wedge to make setup quick and convenient.

Specifications

Product Line PowerEase
Quantity 1 pack
Shipping Condition Room temperature