Learn More
Supelco Powerbank
Catalog No. 1526110001
Change view
Click to view available options
Product Type: Powerbank
Catalog No.
1526110001 Product Type: Powerbank
1 options
Catalog No. 1526110001 Supplier Supelco Supplier No. 1.52611.0001
The Powerbank can be used in order to remotely operate the TLC Explorer Documentation System. The TLC Explorer Documentation System is designed to document and evaluate thin-layer chromatographic (TLC) plates.
  • The powerbank can be used in order to remotely operate the TLC Explorer Documentation System (Cat. No. 1.52610.0001 ).
  • The TLC Explorer Documentation System is designed to document and evaluate thin-layer chromatographic (TLC) plates.
  • It offers a safe, fast and flexible way to visually inspect and digitally record images of TLC plates after their chromatographic development workflow in the laboratory.
  • For detecting different substance classes, the device contains three different LED light sources (white light (VIS), UVA (366 nm) and UVC (254 nm), which illuminate the plates from above in direct-light mode.
  • Recorded plate images can be evaluated to generate reports or export the data for further processing with other systems.
Specifications
Product Type Powerbank