Learn More
DWK Life Sciences Wheaton™ Power Accessories for Socorex™ Acura Electro Electronic Micropipettes
Catalog No. 1370975
Change view
Click to view available options
Type:
Charging Cord
Charging Stand
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
1370975 Charging Stand
1370978 Charging Cord
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1370975 Supplier DWK Life Sciences Supplier No. W820019
Stand allows charging and storing of up to three electronic pipettes or battery packs simultaneously.
  • Charging Stand for Pipettes or Batteries with 3 Positions
Specifications
Number of Channels 3
Type Charging Stand
For Use With (Equipment) Socorex™ Acura™ Electro™ electronic micropipette
Includes Red/Green Charging LED
Voltage 4.8 V