missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PowderTainer™ II 1 to 25 kg Single-Use BPC for powder applications, 5 kg
GREENER_CHOICE

Catalog No. SH3073702
Change view
Click to view available options
Capacity (Metric):
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SH3073702 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3073702 Supplier Thermo Scientific™ Supplier No. SH30737.02

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The 5 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 film that includes 2 ports (1 wash down line, 1 powder port).

The 5 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 film that includes 2 ports (1 wash down line, 1 powder port). The Powdertainer BPC is specifically designed for powder containment and discharge applications, and maintains a closed system for maximum recovery of powder while minimizing the risk of cross contamination.

Features:

  • Three-inch tri-clamp port designed for secure connection to, and easy integration, with existing formulation vessels.
  • Powdertainer BPC features suspensor handle for support during discharge, as well as neck clamp to retain powder prior to discharge.
  • Use this system to eliminate post-use cleaning steps required with re-useable containers.

Applications of the Thermo Scientific™ Powdertainer BPC include:

  • Storage and delivery of powder culture media and buffers.
  • Storage and dispensing of pre-weighed chemicals and other process powders.

Specifications

Includes 2 Ports (1 Wash Down Line, 1 Powder Port)
Film Description CX3-9
Port Detail Powder port: 7.6 cm (3 in.) tri-clamp with integrated seal
Product Line PowderTainer™ II
Dimensions (L x W) (22.1 x 14.3 in.) 56.1 x 36.3 cm
Width (English) 14.3 in.
Product Type Bioprocess Container for Powder
Capacity (Metric) 5 kg
Length (Metric) 56.1 cm
Width (Metric) 36.3 cm