Learn More
Thermo Scientific™ PowderTainer™ II 1 to 25 kg Single-Use BPC for powder applications, 1 kg
The 1 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 film that includes 2 ports (1 wash down line, 1 powder port).
Supplier: Thermo Scientific™ SH3073701
Description
The 1 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 film that includes 2 ports (1 wash down line, 1 powder port). The Powdertainer BPC is specifically designed for powder containment and discharge applications, and maintains a closed system for maximum recovery of powder while minimizing the risk of cross contamination.
Features:
- Three-inch tri-clamp port designed for secure connection to, and easy integration, with existing formulation vessels.
- Powdertainer BPC features suspensor handle for support during discharge, as well as neck clamp to retain powder prior to discharge.
- Use this system to eliminate post-use cleaning steps required with re-useable containers.
Applications of the Thermo Scientific Powdertainer BPC include:
- Storage and delivery of powder culture media and buffers.
- Storage and dispensing of pre-weighed chemicals and other process powders.
Specifications
| 1 kg | |
| 2 Ports (1 Wash Down Line, 1 Powder Port) | |
| CX3-9 | |
| PowderTainer™ II | |
| Bioprocess Container for Powder |
| Powder Application | |
| Silicone | |
| Powder port: 7.6 cm (3 in.) tri-clamp with integrated seal | |
| 16.8 x 12.5 in. (42.7 x 31.8 cm) |