missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PowderTainer™ II 1 to 25kg Single-Use BPC for powder applications, 1 kg
GREENER_CHOICE

Catalog No. SH3086401
Change view
Click to view available options
Capacity (Metric):
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SH3086401 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3086401 Supplier Thermo Scientific™ Supplier No. SH30864.01

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A 2D powder handling system composed of CX3-9 film that includes 1 port (1 powder port), specifically designed for powder containment and discharge applications.

  • A 2D powder handling system composed of CX3-9 film that includes 1 port (1 powder port)
  • Specifically designed for powder containment and discharge applications
  • Maintains a closed system for maximum recovery of powder while minimizing the risk of cross contamination

Features

  • 3 in. tri-clamp port designed for secure connection to, and easy integration with existing formulation vessels
  • Powdertainer BPC features suspensor handle for support during discharge, as well as neck clamp to retain powder prior to discharge
  • Use this system to eliminate post-use cleaning steps required with re-useable containers

Applications

  • Storage and delivery of powder culture media and buffers
  • Storage and dispensing of pre-weighed chemicals and other process powders

Specifications

Includes 1 Port (1 Powder Port)
Film Description CX3-9
Outer Packaging Packages are placed in strong cardboard cartons with product and lot identification.
Port Detail Powder Port: 3 in.
Product Line PowderTainer™ II
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area.
Sterilization Method Non-Irradiated
Tubing Line 1 Washdown Line: N/A
Tubing Line 2 Powder port: 7.6 cm (3 in.) tri-clamp with intergrated seal
Product Type Bioprocess Container for Powder
Capacity (Metric) 1 kg
Length (Metric) 42.7 cm
Width (Metric) 31.8 cm
Show More Show Less