missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Powder Funnel with Flattened Side
GREENER_CHOICE

Catalog No. 31501476
Change view
Click to view available options
Diameter (Metric)Top:
50 mm
75 mm
100 mm
Joint Number:
14/20
19/22
24/40
29/42
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)Top Joint Number
31501476 75 mm 29/42
31501473 50 mm 14/20
31501474 50 mm 19/22
31501475 75 mm 24/40
31501477 100 mm 24/40
31501478 100 mm 29/42
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 31501476 Supplier Synthware™ Supplier No. F192975

Funnels have a 60° side angle and a standard taper inner joint, One side is flattened to allow use with multi-neck flasks.

Specifications

Diameter (Metric)Top 75 mm
Material Borosilicate Glass
Color Clear
For Use With (Application) Organic Synthesis
Joint Number 29/42
Quantity 1 Ea.
Product Type Filling Funnel (Powders)