missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Powder Funnel with Flattened Side

Specifications
Specifications
| Diameter (Metric)Top | 50 mm |
| Material | Borosilicate Glass |
| Color | Clear |
| For Use With (Application) | Organic Synthesis |
| Joint Number | 14/20 |
| Quantity | 1 Ea. |
| Product Type | Filling Funnel (Powders) |