Learn More
Eisco™ Plastic Powder Funnels
Eisco™ Plastic Powder Funnels are an ideal method for transferring powder reagents from one vessel to another.
Supplier:  Eisco™ CH0457D
 View more versions of this product
Catalog No. S24519
Only null left
Explore available promotions
Description
Description
  • Polypropylene powder funnel
  • Designed for transferring powder reagents
  • Translucent
  • Durable for student use
  • Easy to clean
  • Autoclavable
  • Chemical, acid, and alkali resistant
Specifications
Specifications
150 mm
Polypropylene
Autoclavable
Transferring powdered reagents
Filling Funnel (Powders)
700 mL
10.16 cm
Translucent
Parallel stem
Product Suggestions
Product Suggestions