missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Plastic Powder Funnels
Eisco™ Plastic Powder Funnels are an ideal method for transferring powder reagents from one vessel to another.
Supplier: Eisco™ CH0457A
Description
- Polypropylene powder funnel
- Designed for transferring powder reagents
- Translucent
- Durable for student use
- Easy to clean
- Autoclavable
- Chemical, acid, and alkali resistant
Specifications
| 65 mm | |
| Polypropylene | |
| Autoclavable | |
| Transferring powdered reagents | |
| Filling Funnel (Powders) |
| 148.7 mL | |
| 10.16 cm | |
| Translucent | |
| Parallel stem |