missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Plastic Powder Funnels
Description
- Polypropylene powder funnel
- Designed for transferring powder reagents
- Translucent
- Durable for student use
- Easy to clean
- Autoclavable
- Chemical, acid, and alkali resistant
Warranty
1 year limited manufacturer warranty
Specifications
Specifications
| Diameter (Metric)Top | 100 mm |
| Capacity (Metric) | 300 mL |
| Material | Polypropylene |
| Height (Metric) | 7.6 cm |
| Autoclavable | Autoclavable |
| Color | Clear |
| Graduated | No |
| For Use With (Application) | Transferring powdered reagents |
| Includes | Parallel stem |
| Quantity | 1 Ea. |
| Show More |