missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Powder-Coated Three-Prong Retort Clamp

Catalog No. S14089
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S14089 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S14089 Supplier Eisco™ Supplier No. CH0684

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Equipped with diecast jaw with clamping tommy-bar screwthrough swivels.

  • PVC coated
  • Mounted on rod

Specifications

Type 3-Prong Clamp
Material Laboral, PVC
Includes Clamping tommy-bar screwthrough swivels
For Use With (Equipment) Mounted on 13.5cm rod of 8mm dia.
Description 90mm jaw capacity
Length (Metric) 25 cm
Size L x W x H: 11.8 x 5.9 x 3.1 in.
Color Gray
Jaw Type 3-Prong