missing translation for 'onlineSavingsMsg'
Learn More

Povidone K-30, USP, Spectrum™ Chemical

Catalog No. p-7158492
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Amber Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605746 5 g Amber Glass Bottle
18605748 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18605746 Supplier Spectrum Chemical Mfg Cor Supplier No. P14545GM
Only null left

P1454, 9003-39-8, (C6H9NO)n

Spectrum™ Chemical Povidone K-30, USP is used as a binder in pharmaceutical tablets. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

CAS Max % 1
pH 3.0 to 5.0
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 5 g
Grade USP