missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Povidone K-30, USP, Spectrum™ Chemical
Description
Spectrum™ Chemical Povidone K-30, USP is used as a binder in pharmaceutical tablets. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| pH | 3.0 to 5.0 |
| Ignition Residue | 0.001 |
| Packaging | Amber Glass Bottle |
| Quantity | 5 g |
| Grade | USP |