missing translation for 'onlineSavingsMsg'
Learn More

Povidone K-30, Spectrum™ Chemical

Catalog No. 18612940
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612940 500 g Poly Bottle
18612938 100 g Poly Bottle
18612939 5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612940 Supplier Spectrum Chemical Mfg Cor Supplier No. PO147500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9003-39-8

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 500 g
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Povidone K-30

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.