missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Potato Dextrose Agar
Description
Potato Dextrose Agar is used for the isolation, enumeration and identification of yeast and molds. It may also be used to induce sporulation in many fungi.
Specifications
Specifications
| Content And Storage | Room Temperature |
| Description | Potato Dextrose Agar |
| Media Type | Yeast Media |
| Quantity | 500 g |