missing translation for 'onlineSavingsMsg'
Learn More

Potassium Vinyltrifluoroborate 97.0+%, TCI America™

Supplier:  TCI America P147925G

 View more versions of this product

Catalog No. P147925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Vinyltrifluoroborate
White-Yellow
25 g
ZCUMGICZWDOJEM-UHFFFAOYSA-N
potassium;ethenyl(trifluoro)boranuide
23679353
≥97.0% (W)
13682-77-4
C2H3BF3K
potassium vinyltrifluoroborate, potassium trifluoro vinyl borate, potassium ethenyltrifluoroboranuide, vinyltrifluoroboric acid potassium salt, potassium vinyltrifluorborate, potassium ethenyl trifluoroborate, potassium ethenyltrifluoroborate, potassium trifluoro vinyl boranuide, pubchem11308, potassiumvinyltrifluoroborate
[B-](C=C)(F)(F)F.[K+]
133.95
133.95
Crystalline Powder

Chemical Identifiers

13682-77-4
133.95
potassium vinyltrifluoroborate, potassium trifluoro vinyl borate, potassium ethenyltrifluoroboranuide, vinyltrifluoroboric acid potassium salt, potassium vinyltrifluorborate, potassium ethenyl trifluoroborate, potassium ethenyltrifluoroborate, potassium trifluoro vinyl boranuide, pubchem11308, potassiumvinyltrifluoroborate
potassium;ethenyl(trifluoro)boranuide
C2H3BF3K
ZCUMGICZWDOJEM-UHFFFAOYSA-N
23679353
[B-](C=C)(F)(F)F.[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions