missing translation for 'onlineSavingsMsg'
Learn More

Potassium Vinyltrifluoroborate 97.0+%, TCI America™

Catalog No. P14791G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14791G 1 g
P14795G 5 g
P147925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P14791G Supplier TCI America Supplier No. P14791G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13682-77-4
Molecular Formula C2H3BF3K
Molecular Weight (g/mol) 133.95
InChI Key ZCUMGICZWDOJEM-UHFFFAOYSA-N
Synonym potassium vinyltrifluoroborate, potassium trifluoro vinyl borate, potassium ethenyltrifluoroboranuide, vinyltrifluoroboric acid potassium salt, potassium vinyltrifluorborate, potassium ethenyl trifluoroborate, potassium ethenyltrifluoroborate, potassium trifluoro vinyl boranuide, pubchem11308, potassiumvinyltrifluoroborate
PubChem CID 23679353
IUPAC Name potassium;ethenyl(trifluoro)boranuide
SMILES [B-](C=C)(F)(F)F.[K+]

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 133.95
Percent Purity ≥97.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Vinyltrifluoroborate