Learn More
Potassium vinyltrifluoroborate, 97%
CAS: 13682-77-4 | C2H3BF3K | 133.95 g/mol
Supplier: Thermo Scientific Chemicals L1797006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Potassium vinyltrifluoroborate | |
| 230°C (decomposition) | |
| MFCD02093335 | |
| 7777570 | |
| potassium vinyltrifluoroborate, potassium trifluoro vinyl borate, potassium ethenyltrifluoroboranuide, vinyltrifluoroboric acid potassium salt, potassium vinyltrifluorborate, potassium ethenyl trifluoroborate, potassium ethenyltrifluoroborate, potassium trifluoro vinyl boranuide, pubchem11308, potassiumvinyltrifluoroborate | |
| [B-](C=C)(F)(F)F.[K+] | |
| 133.95 | |
| 133.95 |
| 13682-77-4 | |
| C2H3BF3K | |
| 5 g | |
| Hygroscopic | |
| ZCUMGICZWDOJEM-UHFFFAOYSA-N | |
| potassium;ethenyl(trifluoro)boranuide | |
| 23679353 | |
| 97% |
Chemical Identifiers
| 13682-77-4 | |
| 133.95 | |
| ZCUMGICZWDOJEM-UHFFFAOYSA-N | |
| 23679353 | |
| [B-](C=C)(F)(F)F.[K+] |
| C2H3BF3K | |
| MFCD02093335 | |
| potassium vinyltrifluoroborate, potassium trifluoro vinyl borate, potassium ethenyltrifluoroboranuide, vinyltrifluoroboric acid potassium salt, potassium vinyltrifluorborate, potassium ethenyl trifluoroborate, potassium ethenyltrifluoroborate, potassium trifluoro vinyl boranuide, pubchem11308, potassiumvinyltrifluoroborate | |
| potassium;ethenyl(trifluoro)boranuide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only