missing translation for 'onlineSavingsMsg'
Learn More

Potassium vinyltrifluoroborate, 97%

Catalog No. AAL1797003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1797003 1 g
AAL1797006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1797003 Supplier Thermo Scientific Chemicals Supplier No. L1797003

CAS: 13682-77-4 | C2H3BF3K | 133.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13682-77-4
Molecular Formula C2H3BF3K
Molecular Weight (g/mol) 133.95
MDL Number MFCD02093335
InChI Key ZCUMGICZWDOJEM-UHFFFAOYSA-N
Synonym potassium vinyltrifluoroborate, potassium trifluoro vinyl borate, potassium ethenyltrifluoroboranuide, vinyltrifluoroboric acid potassium salt, potassium vinyltrifluorborate, potassium ethenyl trifluoroborate, potassium ethenyltrifluoroborate, potassium trifluoro vinyl boranuide, pubchem11308, potassiumvinyltrifluoroborate
PubChem CID 23679353
IUPAC Name potassium;ethenyl(trifluoro)boranuide
SMILES [B-](C=C)(F)(F)F.[K+]

Specifications

Melting Point 230°C (decomposition)
Quantity 1 g
Beilstein 7777570
Sensitivity Hygroscopic
Formula Weight 133.95
Percent Purity 97%
Chemical Name or Material Potassium vinyltrifluoroborate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only