missing translation for 'onlineSavingsMsg'
Learn More

Potassium vinyltrifluoroborate, 95%

CAS: 13682-77-4 | C2H3BF3K | 133.95 g/mol

$288.29 - $288.29

Chemical Identifiers

CAS 13682-77-4
Molecular Formula C2H3BF3K
Molecular Weight (g/mol) 133.95
MDL Number MFCD02093335
InChI Key ZCUMGICZWDOJEM-UHFFFAOYSA-N
Synonym potassium vinyltrifluoroborate, potassium trifluoro vinyl borate, potassium ethenyltrifluoroboranuide, vinyltrifluoroboric acid potassium salt, potassium vinyltrifluorborate, potassium ethenyl trifluoroborate, potassium ethenyltrifluoroborate, potassium trifluoro vinyl boranuide, pubchem11308, potassiumvinyltrifluoroborate
PubChem CID 23679353
IUPAC Name potassium;ethenyl(trifluoro)boranuide
SMILES [B-](C=C)(F)(F)F.[K+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC387700050
SDS View Documents
Thermo Scientific Chemicals
387700050
5 g Glass bottle
Each for $288.29
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13682-77-4
133.95
ZCUMGICZWDOJEM-UHFFFAOYSA-N
23679353
[B-](C=C)(F)(F)F.[K+]
C2H3BF3K
MFCD02093335
potassium vinyltrifluoroborate, potassium trifluoro vinyl borate, potassium ethenyltrifluoroboranuide, vinyltrifluoroboric acid potassium salt, potassium vinyltrifluorborate, potassium ethenyl trifluoroborate, potassium ethenyltrifluoroborate, potassium trifluoro vinyl boranuide, pubchem11308, potassiumvinyltrifluoroborate
potassium;ethenyl(trifluoro)boranuide

Specifications

13682-77-4
White to Yellow
94% min
C2H3BF3K
5 g
ZCUMGICZWDOJEM-UHFFFAOYSA-N
potassium;ethenyl(trifluoro)boranuide
23679353
95%
Powder
240°C to 242°C
Authentic
Glass bottle
MFCD02093335
potassium vinyltrifluoroborate, potassium trifluoro vinyl borate, potassium ethenyltrifluoroboranuide, vinyltrifluoroboric acid potassium salt, potassium vinyltrifluorborate, potassium ethenyl trifluoroborate, potassium ethenyltrifluoroborate, potassium trifluoro vinyl boranuide, pubchem11308, potassiumvinyltrifluoroborate
[B-](C=C)(F)(F)F.[K+]
133.95
133.95
Technical
Potassium vinyltrifluoroborate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only