missing translation for 'onlineSavingsMsg'
Learn More

Potassium Tris(1-pyrazolyl)borohydride 97.0+%, TCI America™

Catalog No. p-7135138
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14401G 1 g
P14405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14401G Supplier TCI America Supplier No. P14401G
Only null left

Chemical Identifiers

CAS 18583-60-3
Molecular Formula C9H10BKN6
Molecular Weight (g/mol) 252.13
MDL Number MFCD00040346
InChI Key IBNGZWIUPDMZMG-UHFFFAOYSA-N
Synonym potassium hydrotris 1-pyrazolyl borate, potassium hydridotri 1h-pyrazol-1-yl borate 1-, potassium hydro-tris 1-pyrazolyl borate, potassium tri 1h-pyrazol-1-yl hydroborate, potassium tri 1-pyrazolyl borohydride, potassium hydrotris 1-pyrazolato borate, hydrate
IUPAC Name potassium tris(1H-pyrazol-1-yl)boranuide
SMILES [K+].C1=CN(N=C1)[BH-](N1C=CC=N1)N1C=CC=N1

Specifications

Color White
Quantity 1 g
Formula Weight 252.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Tris(1-pyrazolyl)borohydride
TSCA No
Recommended Storage Refrigerator