missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Tripolyphosphate, Spectrum™ Chemical
P1669, 13845-36-8, K5P3O10
$266.10 - $5507.33
Chemical Identifiers
| CAS | 13845-36-8 |
|---|---|
| Molecular Formula | K5O10P3 |
| Molecular Weight (g/mol) | 448.40 |
| InChI Key | ATGAWOHQWWULNK-UHFFFAOYSA-I |
| IUPAC Name | pentapotassium bis(phosphonatooxy)phosphinate |
| SMILES | [K+].[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605741
|
Spectrum Chemical Mfg Cor
P1669500GM |
500 g | Poly Bottle |
Each for $266.10
|
|
||||
|
18605742
|
Spectrum Chemical Mfg Cor
P166925KG |
2.5 kg | Poly Pail |
Each for $865.07
|
|
||||
|
18605743
|
Spectrum Chemical Mfg Cor
P166912KG |
12 kg | Poly Pail |
Each for $2,219.27
|
|
||||
|
18605744
|
Spectrum Chemical Mfg Cor
P166945KGBL |
45 kg | Fiber Drum |
Each for $5,507.33
|
|
||||
Description
Spectrum™ Chemical Potassium TripolyphosphateChemical Identifiers
| 13845-36-8 | |
| 448.40 | |
| pentapotassium bis(phosphonatooxy)phosphinate |
| K5O10P3 | |
| ATGAWOHQWWULNK-UHFFFAOYSA-I | |
| [K+].[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O |
Specifications
| 13845-36-8 | |
| 500 g | |
| ATGAWOHQWWULNK-UHFFFAOYSA-I | |
| pentapotassium bis(phosphonatooxy)phosphinate | |
| Reagent |
| 1 | |
| K5O10P3 | |
| [K+].[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O | |
| 448.40 | |
| Poly Bottle |