missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Trihydrogen Dioxalate Dihydrate 99.0+%, TCI America™
Supplier: TCI America P047525G
Specifications
| Potassium Trihydrogen Dioxalate Dihydrate | |
| White | |
| MFCD00150443,MFCD00150443 | |
| 2811 | |
| GANDVAJEIJXBQJ-UHFFFAOYSA-M | |
| potassium oxalic acid hydrogen oxalate | |
| 131698588 | |
| ≥99.0% (T) |
| 6100-20-5 | |
| C4H3KO8 | |
| 25 g | |
| potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate | |
| [K+].OC(=O)C(O)=O.OC(=O)C([O-])=O | |
| 218.16 | |
| 218.16 | |
| Crystalline Powder |
Chemical Identifiers
| 6100-20-5 | |
| 218.16 | |
| GANDVAJEIJXBQJ-UHFFFAOYSA-M | |
| 131698588 | |
| [K+].OC(=O)C(O)=O.OC(=O)C([O-])=O |
| C4H3KO8 | |
| MFCD00150443,MFCD00150443 | |
| potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate | |
| potassium oxalic acid hydrogen oxalate |
Safety and Handling
EINECSNumber : (2)-0922
TSCA : Yes