missing translation for 'onlineSavingsMsg'
Learn More

Potassium Trihydrogen Dioxalate Dihydrate 99.0+%, TCI America™

Catalog No. P0475500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0475500G 500 g
P047525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0475500G Supplier TCI America Supplier No. P0475500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6100-20-5
Molecular Formula C4H3KO8
Molecular Weight (g/mol) 218.16
MDL Number MFCD00150443,MFCD00150443
InChI Key GANDVAJEIJXBQJ-UHFFFAOYSA-M
Synonym potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate
PubChem CID 131698588
IUPAC Name potassium oxalic acid hydrogen oxalate
SMILES [K+].OC(=O)C(O)=O.OC(=O)C([O-])=O

Specifications

Color White
Quantity 500 g
UN Number 2811
Formula Weight 218.16
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Trihydrogen Dioxalate Dihydrate
EINECSNumber (2)-0922
TSCA Yes