missing translation for 'onlineSavingsMsg'
Learn More

Potassium trihydrogen dioxalate dihydrate, 98+%

CAS: 6100-20-5 | C4H3KO8 | 218.16 g/mol

Supplier:  Thermo Scientific Chemicals A149330I

 View more versions of this product

Catalog No. AAA149330I

Only null left
Explore available promotions

Description

Description

Potassium trihydrogen dioxalate dihydrat is used in metal polishing determination of pH. And it is also used in photography, marble grinding, and to remove ink stains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium trihydrogen dioxalate dihydrat is used in metal polishing determination of pH. And it is also used in photography, marble grinding, and to remove ink stains.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Potassium trihydrogen dioxalate dihydrate
C4H3KO8
5000 g
14,7689
Soluble in water
[K+].OC(=O)C(O)=O.OC(=O)C([O-])=O
218.16
254.19 (218.16 Anhydrous)
6100-20-5
MFCD00150443,MFCD00150443
UN3288
potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate
GANDVAJEIJXBQJ-UHFFFAOYSA-M
potassium;hydron;oxalate;dihydrate
131698588
≥98%

Chemical Identifiers

6100-20-5
218.16
GANDVAJEIJXBQJ-UHFFFAOYSA-M
131698588
C4H3KO8
MFCD00150443,MFCD00150443
potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate
[K+].OC(=O)C(O)=O.OC(=O)C([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 204-874-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only