Learn More
Potassium trihydrogen dioxalate dihydrate, 98+%
CAS: 6100-20-5 | C4H3KO8 | 218.16 g/mol
Supplier: Thermo Scientific Chemicals A149330B
Description
Potassium trihydrogen dioxalate dihydrat is used in metal polishing determination of pH. And it is also used in photography, marble grinding, and to remove ink stains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium trihydrogen dioxalate dihydrat is used in metal polishing determination of pH. And it is also used in photography, marble grinding, and to remove ink stains.
Solubility
Soluble in water
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| Potassium trihydrogen dioxalate dihydrate | |
| C4H3KO8 | |
| 1000 g | |
| 14,7689 | |
| Soluble in water | |
| [K+].OC(=O)C(O)=O.OC(=O)C([O-])=O | |
| 218.16 | |
| 254.19 (218.16 Anhydrous) |
| 6100-20-5 | |
| MFCD00150443,MFCD00150443 | |
| UN3288 | |
| potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate | |
| GANDVAJEIJXBQJ-UHFFFAOYSA-M | |
| potassium;hydron;oxalate;dihydrate | |
| 131698588 | |
| ≥98% |
Chemical Identifiers
| 6100-20-5 | |
| 218.16 | |
| GANDVAJEIJXBQJ-UHFFFAOYSA-M | |
| 131698588 |
| C4H3KO8 | |
| MFCD00150443,MFCD00150443 | |
| potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate | |
| [K+].OC(=O)C(O)=O.OC(=O)C([O-])=O |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber : 204-874-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only