missing translation for 'onlineSavingsMsg'
Learn More

Potassium Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. T154925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T154925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T154925G Supplier TCI America Supplier No. T154925G

Chemical Identifiers

CAS 2926-27-4
Molecular Formula CF3KO3S
Molecular Weight (g/mol) 188.162
MDL Number MFCD00042370
InChI Key GLGXXYFYZWQGEL-UHFFFAOYSA-M
Synonym potassium trifluoromethanesulfonate, potassium triflate, trifluoromethanesulfonic acid potassium salt, potassium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, potassium salt, potassium ion triflate, cf3so3k, acmc-209h7u, potassium trifluoromethanesulfonate salt, potassium tris fluoranyl methanesulfonate
PubChem CID 23664779
IUPAC Name potassium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].[K+]

Specifications

Melting Point 235°C
Color White
Quantity 25 g
Formula Weight 188.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Trifluoromethanesulfonate