missing translation for 'onlineSavingsMsg'
Learn More

Potassium trifluoroacetate, 98%

Catalog No. AAA1739814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1739814 25 g
AAA1739822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1739814 Supplier Thermo Scientific Chemicals Supplier No. A1739814

CAS: 2923-16-2 | C2F3KO2 | 152.11 g/mol

It is a reagent for preparation of aryl trifluoromethyl sulfides, by heating with the diaryl disulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reagent for preparation of aryl trifluoromethyl sulfides, by heating with the diaryl disulfide.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, acids.

Chemical Identifiers

CAS 2923-16-2
Molecular Formula C2F3KO2
Molecular Weight (g/mol) 152.11
MDL Number MFCD00013215
InChI Key CUNPJFGIODEJLQ-UHFFFAOYSA-M
Synonym potassium trifluoroacetate, potassium 2,2,2-trifluoroacetate, trifluoroacetic acid potassium salt, acetic acid, trifluoro-, potassium salt, potassiumtrifluoroacetate, pubchem12605, potassium ion trifluoroacetate, ksc492g5t, trifluoroacetic acid, potassium salt, potassium 2,2,2-tris fluoranyl ethanoate
PubChem CID 23662811
SMILES [K+].[O-]C(=O)C(F)(F)F

Specifications

Melting Point 142°C to 145°C
pH 7
Quantity 25 g
UN Number UN2811
Beilstein 3717603
Sensitivity Hygroscopic
Solubility Information Soluble in water.
IUPAC Name potassium;2,2,2-trifluoroacetate
Formula Weight 152.12
Percent Purity 98%
Chemical Name or Material Potassium trifluoroacetate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 220-877-5
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only