missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiobenzoate, 95%

Catalog No. AAH61741MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61741MD 250 mg
AAH6174103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61741MD Supplier Thermo Scientific Chemicals Supplier No. H61741MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 28170-13-0 | C7H6KOS | 177.28 g/mol

Potassium thiobenzoate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium thiobenzoate is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 28170-13-0
Molecular Formula C7H6KOS
Molecular Weight (g/mol) 177.28
MDL Number MFCD28399074
InChI Key LSBMSRUYNGEWKI-UHFFFAOYSA-N
Synonym benzenecarbothioic acid potassium
PubChem CID 87087528
IUPAC Name benzenecarbothioic S-acid;potassium
SMILES [K].SC(=O)C1=CC=CC=C1

Specifications

Physical Form Solid
Quantity 250 mg
Formula Weight 176.28
Percent Purity 95%
Chemical Name or Material Potassium thiobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only