missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetraphenylborate, 97%

Catalog No. p-7052681
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5527606 5 g
AAH5527614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5527606 Supplier Thermo Scientific Chemicals Supplier No. H5527606

CAS: 3244-41-5 | C24H20BK | 358.332 g/mol

It is an active pharmaceutical intemediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intemediate.

Solubility
Low solubility in water. Soluble in organic solvents.

Notes
Desiccate at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 3244-41-5
Molecular Formula C24H20BK
Molecular Weight (g/mol) 358.332
MDL Number MFCD03093877
InChI Key MAMCUZQNCMYPCY-UHFFFAOYSA-N
Synonym potassium tetraphenylborate, potassium tetraphenylboron, potassium tetraphenylboride, potassium tetrakis phenyl borate, potassium tetraphenyl boron, potassium tetraphenylborate 97, potassium tetraphenylborate 1-
PubChem CID 23665558
IUPAC Name potassium;tetraphenylboranuide
SMILES [B-](C1=CC=CC=C1)(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[K+]

Specifications

Melting Point >300°C
Color White
Physical Form Powder
Quantity 5 g
Linear Formula KB(C6H5)4
Solubility Information Low solubility in water. Soluble in organic solvents.
Formula Weight 358.33
Percent Purity 97%
Chemical Name or Material Potassium tetraphenylborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only