missing translation for 'onlineSavingsMsg'
Learn More

Potassium Tetraoxalate, 99%, Spectrum™ Chemical

Catalog No. 18612916
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612916 25 g Poly Bottle
18612918 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18612916 Supplier Spectrum Chemical Mfg Cor Supplier No. P141825GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 6100-20-5

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.