missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium tetranitroplatinate(II), Pt 42.6% min
CAS: 13815-39-9 | K2N4O8Pt | 457.30 g/mol
$248.90 - $972.68
Chemical Identifiers
| CAS | 13815-39-9 |
|---|---|
| Molecular Formula | K2N4O8Pt |
| Molecular Weight (g/mol) | 457.30 |
| MDL Number | MFCD00011375 |
| InChI Key | GQUAVGOGNMMUFS-UHFFFAOYSA-J |
| Synonym | potassium tetranitroplatinate ii, platinum potassium nitrite, dipotassium ion tetranitroplatinumdiuide, dipotassium tetranitroplatinumdiuide |
| PubChem CID | 16211695 |
| SMILES | [K+].[K+].[Pt++].[O-]N=O.[O-]N=O.[O-]N=O.[O-]N=O |
Description
Potassium tetranitroplatinate(II) is used widely as laboratory chemicals and in manufacture of substances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13815-39-9 | |
| 457.30 | |
| GQUAVGOGNMMUFS-UHFFFAOYSA-J | |
| 16211695 |
| K2N4O8Pt | |
| MFCD00011375 | |
| potassium tetranitroplatinate ii, platinum potassium nitrite, dipotassium ion tetranitroplatinumdiuide, dipotassium tetranitroplatinumdiuide | |
| [K+].[K+].[Pt++].[O-]N=O.[O-]N=O.[O-]N=O.[O-]N=O |
Specifications
| 13815-39-9 | |
| 1 g | |
| MFCD00011375 | |
| It is slightly soluble in water. It is soluble in organic or non-aqueous solvent. | |
| [K+].[K+].[Pt++].[O-]N=O.[O-]N=O.[O-]N=O.[O-]N=O | |
| 457.30 | |
| 457.32 | |
| Potassium tetranitroplatinate(II) |
| Crystalline | |
| K2N4O8Pt | |
| potassium tetranitroplatinate ii, platinum potassium nitrite, dipotassium ion tetranitroplatinumdiuide, dipotassium tetranitroplatinumdiuide | |
| GQUAVGOGNMMUFS-UHFFFAOYSA-J | |
| dipotassium;platinum;tetranitrite | |
| 16211695 | |
| Pt 42.6% min. |
Safety and Handling
EINECSNumber : 237-491-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only