missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetracyanopalladate(II) hydrate

Catalog No. AA1053503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1053503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1053503 Supplier Thermo Scientific Chemicals Supplier No. 01053503

CAS: 14516-46-2 | C4K2N4Pd | 288.69 g/mol

Potassium tetracyanopalladate(II) hydrate is used as a primary and secondary intermediate in Chemical & pharmaceutical research. it is also used as catalysts, as precursors of metallic palladium, in preparative chemical production, in photography, in electroplating and medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14516-46-2
Molecular Formula C4K2N4Pd
Molecular Weight (g/mol) 288.69
MDL Number MFCD00150431
InChI Key NKYOTIKKMTWAOI-UHFFFAOYSA-N
IUPAC Name dipotassium palladiumtetracarbonitrile
SMILES [K+].[K+].N#C[Pd](C#N)(C#N)C#N

Specifications

Color White
Physical Form Crystalline
Quantity 1 g
Solubility Information It is slightly soluble in water. It is soluble in organic or non-aqueous solvent.
Formula Weight 288.68 (Anhydrous)
Chemical Name or Material Potassium tetracyanopalladate(II) hydrate
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 238-520-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only