missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrachloroplatinate(II), Premion™, 99.99% (metals basis), Pt 46.4% min

Catalog No. p-7059095
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4394603 1 g
AA4394606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4394603 Supplier Thermo Scientific Chemicals Supplier No. 04394603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10025-99-7 | Cl4H4K2Pt | 419.11 g/mol

Potassium tetrachloroplatinate(II) is used as important reagent for the preparation of other coordination complexes of platinum. It is also used in the induction of apoptosis in human breast cancer cells. Also used in the preparation of camptothecin-linked platinum anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10025-99-7
Molecular Formula Cl4H4K2Pt
Molecular Weight (g/mol) 419.11
MDL Number MFCD00011378
InChI Key JCAXKWJYKUEOFO-UHFFFAOYSA-N
Synonym potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate
PubChem CID 61440
SMILES Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]

Specifications

Melting Point (decomposition)
Color Red to Brown
Physical Form Crystalline
Quantity 1 g
Linear Formula K2PtCl4
UN Number UN3288
Merck Index 14,7682
Solubility Information Soluble in water (9.3g/L). Insoluble in ethyl alcohol.
IUPAC Name dipotassium tetrachloroplatinumdiuide
Formula Weight 415.11
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Pt 46.4% min
Odor Odorless
Density 3.38 g/mL
Assay (metals basis)
Chemical Name or Material Potassium tetrachloroplatinate (II)
Show More Show Less
Hazard Category H301-H315-H317-H318-H334-H335
Hazard Statement GHS H Statement
H301-H334-H318-H290-H315-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May be corrosive to metals.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 233-050-9
RTECSNumber TP1850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only