missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrachloroplatinate(II), 99.9% (metals basis), Pt 46.0% min

Catalog No. p-4824902
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1104803 1 g
AA1104806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1104803 Supplier Thermo Scientific Chemicals Supplier No. 01104803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10025-99-7 | Cl4H4K2Pt | 419.11 g/mol

Potassium tetrachloroplatinate solution is used in the preparation of platinum (Pt) nanoparticles. Ligands like ammonia or triphenyl phosphine can able to replace the chloride ligands to afford various derivatives. The replacement by ammonia results in the preparation of cisplatin, which is used in the cancer treatment. Further, it is used as catalyst in the hydroarylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10025-99-7
Molecular Formula Cl4H4K2Pt
Molecular Weight (g/mol) 419.11
MDL Number MFCD00011378
InChI Key JCAXKWJYKUEOFO-UHFFFAOYSA-N
Synonym potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate
PubChem CID 61440
SMILES Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]

Specifications

Melting Point (decomposition)
Color Red to Brown
Physical Form Crystalline
Quantity 1 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in hot water. Slightly soluble in water.
IUPAC Name dipotassium;tetrachloroplatinum(2-)
Formula Weight 415.11
Odor Odorless
Appearance Red-brown crystals
Density 3.38
Chemical Name or Material Potassium tetrachloroplatinate(II), Pt 46.0% min.
Show More Show Less
Hazard Category H301-H315-H317-H318-H334-H335
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P342+P311-P362-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 233-050-9
RTECSNumber TP1850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only