missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrachloroplatinate, 99.99%, (trace metal basis)

Catalog No. AC437030050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437030050 5 g Glass bottle
AC437030010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437030050 Supplier Thermo Scientific Chemicals Supplier No. 437030050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10025-99-7 | Cl4H4K2Pt | 419.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis

Chemical Identifiers

CAS 10025-99-7
Molecular Formula Cl4H4K2Pt
Molecular Weight (g/mol) 419.11
MDL Number MFCD00011378
InChI Key JCAXKWJYKUEOFO-UHFFFAOYSA-N
Synonym potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate
PubChem CID 61440
IUPAC Name dipotassium tetrachloroplatinumdiuide
SMILES Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]

Specifications

Melting Point 250.0°C
Color Brown or Orange to Red
Physical Form Crystals
Quantity 5 g
Packaging Glass bottle
Assay Percent Range 99.99%
Linear Formula K2PtCl4
Merck Index 15,7801
Solubility Information Solubility in water: 10g/L (20°C)
Formula Weight 415.1
Percent Purity 99.99%
Grade Trace Metal Basis
Chemical Name or Material Potassium tetrachloroplatinate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 233-050-9

RUO – Research Use Only