missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-pentyloxide, 25% w/w in toluene

Catalog No. AAL15111AK
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15111AK 250 mL
AAL15111AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL15111AK Supplier Thermo Scientific Chemicals Supplier No. L15111AK
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 41233-93-6 | C5H11KO | 126.24 g/mol

Potassium tert-pentyloxide is a strong, hindered base used in combination with lithium diisopropyl amide to generate a superbase for the selective metallation of the aralkyl system at the 6-position of protected estradiols. It is used in organic synthesis, agrochemicals, pharmaceuticals, colorants and aroma chemicals. It acts as a catalyst in polymerization and isomerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tert-pentyloxide is a strong, hindered base used in combination with lithium diisopropyl amide to generate a superbase for the selective metallation of the aralkyl system at the 6-position of protected estradiols. It is used in organic synthesis, agrochemicals, pharmaceuticals, colorants and aroma chemicals. It acts as a catalyst in polymerization and isomerizations.

Notes
Air and moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and acids. Reacts violently with water.

Chemical Identifiers

CAS 41233-93-6
Molecular Formula C5H11KO
Molecular Weight (g/mol) 126.24
MDL Number MFCD00064808
InChI Key ZRLVQFQTCMUIRM-UHFFFAOYSA-N
Synonym potassium 2-methylbutan-2-olate, potassium tert-amylate, potassium tert-pentoxide, potassium tert-pentylate, potassium tert-pentoxide solution, potassium 2-methyl-2-butoxide, 2-butanol, 2-methyl-, potassium salt, potassium tert-amyloxide, 2-butanol, 2-methyl-, potassium salt 1:1
PubChem CID 23683543
IUPAC Name potassium;2-methylbutan-2-olate
SMILES [K+].CCC(C)(C)[O-]

Specifications

Density 0.87
Flash Point 4°C (39°F)
Quantity 250 mL
UN Number UN2924
Beilstein 4149025
Sensitivity Air and moisture sensitive
Formula Weight 126.24
Concentration or Composition (by Analyte or Components) 25% w/w in toluene
Chemical Name or Material Potassium tert-pentyloxide
Hazard Category H225-H304-H314-H335-H336-H361d-H373
Hazard Statement GHS H Statement
H225-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 255-272-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only