missing translation for 'onlineSavingsMsg'
Learn More

Potassium [[(tert-Butoxycarbonyl)amino]methyl]trifluoroborate 97.0+%, TCI America™

Catalog No. P21555G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21555G 5 g
P21551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21555G Supplier TCI America Supplier No. P21555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1314538-55-0 | Formula: C6H12BF3KNO2 | Weight: 237.071 g/mol

Chemical Identifiers

CAS 1314538-55-0
Molecular Formula C6H12BF3KNO2
Molecular Weight (g/mol) 237.071
MDL Number MFCD19686142
InChI Key WSOSBCPWIQUNLD-UHFFFAOYSA-N
Synonym Potassium (N-Boc-aminomethyl)trifluoroborate
PubChem CID 53483531
IUPAC Name potassium;trifluoro-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]boranuide
SMILES [B-](CNC(=O)OC(C)(C)C)(F)(F)F.[K+]

Specifications

Melting Point 181°C
Color White
Quantity 5 g
Formula Weight 237.07
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Potassium [[(tert-Butoxycarbonyl)amino]methyl]trifluoroborate