missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-butoxide, pure, 20 wt.% solution in THF

Catalog No. p-4510596
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC364990500 50 mL Glass bottle
AC364992500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC364990500 Supplier Thermo Scientific Chemicals Supplier No. 364990500
Only null left

CAS: 865-47-4 | C4H9KO | 112.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Potassium tert-butoxide
CAS 865-47-4, 109-99-9
Color Amber to Colorless
Density 0.9290g/mL
Flash Point −21°C
Packaging Glass bottle
Molecular Formula C4H9KO
Linear Formula (CH3)3COK
Quantity 50 mL
Fieser 01,911; 02,336; 03,233; 04,399; 05,544; 06,477; 08,407; 09,380; 10,323; 11,432; 12,97; 14,264; 17,289
Specific Gravity 0.929
Synonym potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
Solubility Information Solubility in water: reacts with water.
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES CC(C)(C)[O-].[K+]
IUPAC Name potassium;2-methylpropan-2-olate
Molecular Weight (g/mol) 112.21
PubChem CID 23665647
Formula Weight 112.21
Grade Pure
Physical Form Solution
Show More Show Less

RUO – Research Use Only