missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-butoxide, pure, 1.6-1.7M (20 wt.%) solution in THF, AcroSeal™

Catalog No. p-4089534
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428791000 100 mL AcroSeal™ Glass Bottle
AC428798000 800 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428791000 Supplier Thermo Scientific Chemicals Supplier No. 428791000
Only null left

CAS: 865-47-4 | C4H9KO | 112.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Potassium tert-butoxide
Name Note Pure, 1.7M (20 wt.%) solution in THF
Color Amber to Yellow
CAS 865-47-4, 109-99-9
Physical Form Solution
Quantity 100 mL
Molecular Formula C4H9KO
Linear Formula (CH3)3COK
MDL Number MFCD00012162
Flash Point −21°C
Synonym potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
Solubility Information Solubility in water: reacts with water
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES CC(C)(C)[O-].[K+]
IUPAC Name potassium;2-methylpropan-2-olate
Molecular Weight (g/mol) 112.21
PubChem CID 23665647
Formula Weight 112.21
Grade Pure
Concentration or Composition (by Analyte or Components) 1.60 to 1.75M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.929
Density 0.9290g/mL
Show More Show Less

RUO – Research Use Only