missing translation for 'onlineSavingsMsg'
Learn More

Potassium Tellurite, Powder, Reagent, 90.6%, Spectrum™ Chemical

PO310, 7790-58-1, K2TeO3

Supplier:  Spectrum Chemical Mfg Cor PO310100GM

 View more versions of this product

Catalog No. 18605739


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Potassium Tellurite, Powder, Reagent
Specifications

Specifications

7790-58-1
100 g
BFPJYWDBBLZXOM-UHFFFAOYSA-L
dipotassium tellurite
96%
Amber Glass Bottle
1
K2O3Te
[K+].[K+].[O-][Te]([O-])=O
253.79
Reagent

Chemical Identifiers

7790-58-1
253.79
dipotassium tellurite
K2O3Te
BFPJYWDBBLZXOM-UHFFFAOYSA-L
[K+].[K+].[O-][Te]([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions