missing translation for 'onlineSavingsMsg'
Learn More

Potassium Tellurite, Powder, Reagent, 90.6%, Spectrum™ Chemical

Catalog No. 18605738
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605738 25 g Amber Glass Bottle
18605737 10 g Amber Glass Bottle
18605739 100 g Amber Glass Bottle
18605740 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605738 Supplier Spectrum Chemical Mfg Cor Supplier No. PO31025GM
Only null left

PO310, 7790-58-1, K2TeO3

Spectrum™ Chemical Potassium Tellurite, Powder, Reagent

Chemical Identifiers

CAS 7790-58-1
Molecular Formula K2O3Te
Molecular Weight (g/mol) 253.79
InChI Key BFPJYWDBBLZXOM-UHFFFAOYSA-L
IUPAC Name dipotassium tellurite
SMILES [K+].[K+].[O-][Te]([O-])=O

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 96%
Grade Reagent
Packaging Amber Glass Bottle