missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sulfamate 98.0+%, TCI America™

Catalog No. S0164100G
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0164100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0164100G Supplier TCI America Supplier No. S0164100G
Only null left

Chemical Identifiers

CAS 13823-50-2
Molecular Formula H2KNO3S
Molecular Weight (g/mol) 135.178
MDL Number MFCD00059941
InChI Key BTAAXEFROUUDIL-UHFFFAOYSA-M
Synonym potassium sulfamate, potassium sulphamate, sulfamic acid,potassium salt 1:1, sulfamic acid, monopotassium salt, sulfamic acid, potassium salt 1:1, amidosulfonic acid, potassium salt, potassium amidosulfonate, acmc-209chf, sulfamic acid potassium salt, aminosulfonic acid potassium salt
PubChem CID 23687442
IUPAC Name potassium;sulfamate
SMILES NS(=O)(=O)[O-].[K+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 100 g
Formula Weight 135.18
Percent Purity ≥98.0% (T)
Chemical Name or Material Potassium Sulfamate