missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfamate, 98%

CAS: 13823-50-2 | H2KNO3S | 135.178 g/mol

Supplier:  Thermo Scientific Chemicals B2444722

 View more versions of this product

Catalog No. AAB2444722


Only null left
Explore available promotions

Description

Description

Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications

Specifications

Potassium sulfamate
13823-50-2
100 g
H2NSO3K
potassium sulfamate, potassium sulphamate, sulfamic acid,potassium salt 1:1, sulfamic acid, monopotassium salt, sulfamic acid, potassium salt 1:1, amidosulfonic acid, potassium salt, potassium amidosulfonate, acmc-209chf, sulfamic acid potassium salt, aminosulfonic acid potassium salt
NS(=O)(=O)[O-].[K+]
135.178
135.18
Hygroscopic
White
Crystalline Solid
H2KNO3S
MFCD00059941
BTAAXEFROUUDIL-UHFFFAOYSA-M
potassium;sulfamate
23687442
98%

Chemical Identifiers

13823-50-2
135.178
BTAAXEFROUUDIL-UHFFFAOYSA-M
23687442
NS(=O)(=O)[O-].[K+]
H2KNO3S
MFCD00059941
potassium sulfamate, potassium sulphamate, sulfamic acid,potassium salt 1:1, sulfamic acid, monopotassium salt, sulfamic acid, potassium salt 1:1, amidosulfonic acid, potassium salt, potassium amidosulfonate, acmc-209chf, sulfamic acid potassium salt, aminosulfonic acid potassium salt
potassium;sulfamate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only