missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfamate, 98%

Catalog No. AAB2444736
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2444736 500 g
AAB2444714 25 g
AAB2444722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2444736 Supplier Thermo Scientific Chemicals Supplier No. B2444736
Only null left

CAS: 13823-50-2 | H2KNO3S | 135.178 g/mol

Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium sulfamate is used as a food additive, used in water treatment and as a cleaning agents. It is used in the synthesis of heterocyclic nitramines.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 13823-50-2
Molecular Formula H2KNO3S
Molecular Weight (g/mol) 135.178
MDL Number MFCD00059941
InChI Key BTAAXEFROUUDIL-UHFFFAOYSA-M
Synonym potassium sulfamate, potassium sulphamate, sulfamic acid,potassium salt 1:1, sulfamic acid, monopotassium salt, sulfamic acid, potassium salt 1:1, amidosulfonic acid, potassium salt, potassium amidosulfonate, acmc-209chf, sulfamic acid potassium salt, aminosulfonic acid potassium salt
PubChem CID 23687442
IUPAC Name potassium;sulfamate
SMILES NS(=O)(=O)[O-].[K+]

Specifications

Color White
Physical Form Crystalline Solid
Quantity 500 g
Linear Formula H2NSO3K
Formula Weight 135.18
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Potassium sulfamate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA

RUO – Research Use Only