missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sorbate 99.0+%, TCI America™

Catalog No. p-7135786
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S005725G 25 g
S0057500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S005725G Supplier TCI America Supplier No. S005725G
Only null left

Chemical Identifiers

CAS 24634-61-5
Molecular Formula C6H7KO2
Molecular Weight (g/mol) 150.218
MDL Number MFCD00016546
InChI Key CHHHXKFHOYLYRE-STWYSWDKSA-M
Synonym potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
PubChem CID 23676745
ChEBI CHEBI:77868
IUPAC Name potassium;(2E,4E)-hexa-2,4-dienoate
SMILES CC=CC=CC(=O)[O-].[K+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 150.22
Percent Purity ≥99.0% (T)
Physical Form Granules
Chemical Name or Material Potassium Sorbate
EINECSNumber (2)-1076
RTECSNumber WG2170000
TSCA Yes