missing translation for 'onlineSavingsMsg'
Learn More

Potassium sorbate, 99%

CAS: 24634-61-5 | C6H7KO2 | 150.22 g/mol

Supplier:  Thermo Scientific Chemicals 291740050

Quantity 5 kg
Packaging Plastic drum

 View more versions of this product

Catalog No. AC291740050


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

24634-61-5
150.22
potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
CHEBI:77868
CC=CC=CC(=O)[O-].[K+]
C6H7KO2
CHHHXKFHOYLYRE-STWYSWDKSA-M
23676745
potassium;(2E,4E)-hexa-2,4-dienoate

Specifications

Potassium sorbate
24634-61-5
White to Cream
Authentic
Plastic drum
CH3CH=CHCH=CHCOOK
15, 8847
Solubility in water: 58.2 % (20°C). Other solubilities: soluble in alcohols: 6.5% (20°C)
CC=CC=CC(=O)[O-].[K+]
150.22
CHEBI:77868
99%
Passes test
270.0°C
1% (105°C, 3 hrs) max.
99%
C6H7KO2
5 kg
potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
CHHHXKFHOYLYRE-STWYSWDKSA-M
potassium;(2E,4E)-hexa-2,4-dienoate
23676745
150.22
Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Warning

EINECSNumber : 246-376-1

SDS
Documents

Documents

Promotions

Promotions