missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sodium Tartrate Tetrahydrate, 99.0-102.0%, (ACS Reagent Grade), MP Biomedicals
Supplier: MP Biomedicals Inc 0215256390
Specifications
| Potassium Sodium Tartrate Tetrahydrate | |
| 500 g | |
| C4H12KNaO10 | |
| VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
| potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate | |
| 2724148 | |
| ACS Reagent |
| 6381-59-5 | |
| 99.0-102.0% | |
| potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
| C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] | |
| 282.218 | |
| 282.22 |
Chemical Identifiers
| 6381-59-5 | |
| 282.218 | |
| potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
| potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
| C4H12KNaO10 | |
| VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
| 2724148 | |
| C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
Safety and Handling
Recommended Storage : Room Temperature