missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sodium Tartrate ACS AR Crystal, Macron Fine Chemicals™
macron
$3935.09 - $3935.09
Chemical Identifiers
| CAS | 6381-59-5 |
|---|---|
| Molecular Formula | C4H12KNaO10 |
| Molecular Weight (g/mol) | 282.218 |
| InChI Key | VZOPRCCTKLAGPN-UHFFFAOYSA-L |
| Synonym | potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j |
| PubChem CID | 2724148 |
| IUPAC Name | potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
| SMILES | C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
Chemical Identifiers
| 6381-59-5 | |
| 282.218 | |
| potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
| potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
| C4H12KNaO10 | |
| VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
| 2724148 | |
| C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
Specifications
| 6381-59-5 | |
| C4H12KNaO10 | |
| potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
| C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] | |
| 282.218 | |
| 282.22 | |
| Crystalline |
| POLYSTORMOR | |
| 500 g | |
| VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
| potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate | |
| 2724148 | |
| ACS | |
| Potassium Sodium Tartrate, 4-Hydrate |